Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Sodium triazide

103692KEA 124 GBP
103692K 103694M 10369AS
Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
Melting Pt: 275
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1 II
Merck Index: 13,08653

Order Now

Specification Test Results

Assay Min. 99.0 %
Appearance White crystalline powder
Identification Passes test
Free alkalinity Max. 0.05 meq/g
Insolubility in water Max. 0.015 %
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Cr (Chromium) Max. 20 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR