2-Ethoxyethanol ≥99.5%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve

103426REA 151 GBP
103426R
2-Ethoxyethanol ≥99.5%, AnalaR NORMAPUR® analytical reagent
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
MW: 90.12 g/mol
Boiling Pt: 135
Melting Pt: –70 °C
Density: 0.932 g/cm³ (20 °C)
Flash Pt: 40 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3 III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
IR Spectrum Passes test
Acidity Max. 0.0030 meq/g
Colouration Max. 20 APHA
Density (20/4) 0.929 - 0.930
Evaporation residue Max. 100 ppm
Peroxides (as H2O2) Max. 0.02 %
Water Max. 0.1 %
Al (Aluminium) Max. 10 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 2 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.5 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR