Rubidium ≥99.75% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Breakseal ampoule

10315.03 10315.06 10315.14 10315.22
10315.03EA 120 GBP
10315.03 10315.06 10315.14 10315.22
Rubidium ≥99.75% (metals basis)
Rubidium

Purity: 99.75% CAS No.: 7440-17-7 Physical Form: Vacuum sealed in breakseal ampoules Appearance: Silver-white UN No.: 1423 MDL No.: MFCD00134055

Formula: Rb
Boiling Pt: 700 °C (1013 hPa)
Melting Pt: 39 °C
Density: 0.83 g/cm³ (1399.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134055
CAS Number: 7440-17-7
EINECS: 231-126-6
UN: 1423
ADR: 4.3,I
Merck Index: 12,08439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR