Chlorobenzene 99%, Reagent Grade

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

101389-2.5L
101389-2.5LEA 95.1 GBP
101389-2.5L
Chlorobenzene 99%, Reagent Grade
Chlorobenzene

  • ≥95% for general laboratory use.

Formula: C₆H₅Cl
Boiling Pt: 130…132 °C (1.013 hPa)
Melting Pt: –45 °C
Density: 1.107…1.109 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) min. 98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR