Graphite ≥99% (metals basis), rods, Length: 61 cm (24 in), Ø 6.3 mm (0.25 in).

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10135.KI 10135.KL
10135.KIEA 60.2 GBP
10135.KI 10135.KL
Graphite ≥99% (metals basis), rods, Length: 61 cm (24 in), Ø 6.3 mm (0.25 in).
Graphite

Purity: 99% Physical Form: ≈40g/pc MDL No.: MFCD00144065

Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR