Bovine serum albumin (BSA)

Supplier: SEQENS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1010-30-100ML 1010-30-500ML 1010-30-1L
1010-30-100MLEA 76.4 GBP
1010-30-100ML 1010-30-500ML 1010-30-1L
Bovine serum albumin (BSA)
Proteins and Peptides

SEQENS IVD manufactures Bovine Serum Albumin (BSA) derived from plasma, collected from healthy animals of French origin. This product is obtained by 'Heat Shock' method, which ensures higher purity and preserves BSA intrinsic properties.


  • Albumin purity: >98%
  • Total proteins: >290 g/L
  • Endotoxins: <15,0 Endotoxin Unit/mg dry extract
  • Shelf life: 3 years
  • Sterile filtered 0,2 µm


Full traceability and batch-to-batch repeatability guaranteed.


Delivery information: Supplied as a ready to use 30% solution in deionised water.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR