Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10043.HW 10043.HX 10043.GJ
10043.HWEA 137 GBP
10043.HW 10043.HX 10043.GJ
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Molybdenum

Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈10.2g/100x100mm MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR