Cesium chloride ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

10018.14 10018.22
10018.14EA 52.9 GBP
10018.14 10018.22
Cesium chloride ≥99.9% (metals basis)
Cesium chloride

Purity: 99.9% CAS No.: 7647-17-8 Molecular Formula: CsCl Formula Weight: 168.36 Physical Form: Crystalline Density: 3.99 Refractive Index: 1.6418 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 646° Boiling Point: 1303° Merck Reference: 14,2011 MDL No.: MFCD00010955


For density gradient work to separate and purify proteins

Formula: CsCl
Boiling Pt: 1303 °C
Melting Pt: 646 °C
Density: 3.99
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 14,2011

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR