Boron standard solution, 1000 mg/L B in water (from B(OH₃)) Certipur® standard for ICP, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.70307.0100
1.70307.0100EA 184 GBP
1.70307.0100
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) Certipur® standard for ICP, Supelco®
Standards Cationic Standards Boron Standards
H₃BO₃ in water.
Certifications: Traceable to SRM from NIST. The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Delivery information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.