HPLC columns, Chromolith® CN
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.52046.0001 1.52047.0001 1.52048.0001 1.52050.0001
1.52046.0001EA 511 GBP
1.52046.0001 1.52047.0001 1.52048.0001 1.52050.0001
HPLC columns, Chromolith® CN
Chromatography Columns
Cyano columns are generally more polar than traditional alkyl silica columns. The functional groups are highly ordered, reducing steric hindrance for the solute. The modification also allows cation exchange activity, which is higher at neutral pH than in acidic conditions. Chromolith® CN columns are suitable for the separation of alkaloids, oils, flavonoids, glycols, phenols, phthalates, steroids and sulfonamides.