HPLC columns, LiChrospher®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.50148.0001 1.50149.0001 1.50154.0001 1.50155.0001 1.50156.0001 1.50158.0001 1.50159.0001 1.50316.0001 1.50329.0001 1.50377.0001 1.50477.0001 1.50734.0001 1.50742.0001 1.50820.0001 1.50822.0001 1.50823.0001 1.50824.0001 1.50825.0001 1.50826.0001 1.50827.0001 1.50828.0001 1.50829.0001 1.50830.0001 1.50832.0001 1.50833.0001 1.50834.0001 1.50836.0001 1.50837.0001 1.50838.0001 1.50839.0001 1.50840.0001 1.50842.0001 1.50843.0001 1.50845.0001 1.50847.0001 1.50848.0001 1.50858.0001 1.50892.0001 1.50930.0001 1.50931.0001 1.50936.0001 1.50937.0001 1.50942.0001 1.50943.0001 1.50945.0001 1.50955.0001 1.50956.0001 1.50957.0001 1.50958.0001 1.50959.0001 1.50960.0001 1.50961.0001 1.50962.0001 1.50963.0001 1.50981.0001 1.50982.0001 1.50983.0001 1.50984.0001 1.50987.0001 1.50993.0001 1.50995.0001 1.50549.0001 1.50550.0001 1.50573.0001 1.50574.0001 1.50575.0001 1.50578.0001 1.50579.0001 1.50580.0001 1.50581.0001 1.50582.0001 1.50583.0001 1.50636.0001 1.50547.0001 1.50641.0001 1.50642.0001 1.50545.0001 1.50546.0001 1.50600.0001 1.50601.0001 1.50602.0001 1.50603.0001 1.50604.0001 1.50605.0001 1.50635.0001 1.50638.0001 1.50639.0001 1.50641.1003 1.50823.1003 1.50833.1003 1.50155.1003 1.50600.1003 1.50602.1003 1.50640.1003 1.50154.1003 1.50159.1003 1.50545.1003 1.50377.1003 1.50477.1003 1.50573.1003 1.50574.1003
1.50148.0001EA 403 GBP
1.50148.0001 1.50149.0001 1.50154.0001 1.50155.0001 1.50156.0001 1.50158.0001 1.50159.0001 1.50316.0001 1.50329.0001 1.50377.0001 1.50477.0001 1.50734.0001 1.50742.0001 1.50820.0001 1.50822.0001 1.50823.0001 1.50824.0001 1.50825.0001 1.50826.0001 1.50827.0001 1.50828.0001 1.50829.0001 1.50830.0001 1.50832.0001 1.50833.0001 1.50834.0001 1.50836.0001 1.50837.0001 1.50838.0001 1.50839.0001 1.50840.0001 1.50842.0001 1.50843.0001 1.50845.0001 1.50847.0001 1.50848.0001 1.50858.0001 1.50892.0001 1.50930.0001 1.50931.0001 1.50936.0001 1.50937.0001 1.50942.0001 1.50943.0001 1.50945.0001 1.50955.0001 1.50956.0001 1.50957.0001 1.50958.0001 1.50959.0001 1.50960.0001 1.50961.0001 1.50962.0001 1.50963.0001 1.50981.0001 1.50982.0001 1.50983.0001 1.50984.0001 1.50987.0001 1.50993.0001 1.50995.0001 1.50549.0001 1.50550.0001 1.50573.0001 1.50574.0001 1.50575.0001 1.50578.0001 1.50579.0001 1.50580.0001 1.50581.0001 1.50582.0001 1.50583.0001 1.50636.0001 1.50547.0001 1.50641.0001 1.50642.0001 1.50545.0001 1.50546.0001 1.50600.0001 1.50601.0001 1.50602.0001 1.50603.0001 1.50604.0001 1.50605.0001 1.50635.0001 1.50638.0001 1.50639.0001 1.50641.1003 1.50823.1003 1.50833.1003 1.50155.1003 1.50600.1003 1.50602.1003 1.50640.1003 1.50154.1003 1.50159.1003 1.50545.1003 1.50377.1003 1.50477.1003 1.50573.1003 1.50574.1003
HPLC columns, LiChrospher®
Chromatography Columns
LiChrospher® is Merck's reliable and versatile, traditionally-produced, spherical silica. LiChrospher® is available with different modifications.
LiChrospher® Si 60 and Si 100 are versatile HPLC spherical silica sorbents for normal phase chromatography.
LiChrospher® CN has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. In addition, selective charged interactions makes it even more versatile.
LiChrospher® NH₂ possessesMore Product Information