Polyvinyl alcohol, M.W. 115,000 88, EMPROVE® EXPERT JPE, Ph. Eur., USP excipient_suitable, hydrolysed, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

1.41351.1000 1.41351.9029
1.41351.1000EA 522 GBP
1.41351.1000 1.41351.9029
Polyvinyl alcohol, M.W. 115,000 88, EMPROVE® EXPERT JPE, Ph. Eur., USP excipient_suitable, hydrolysed, SAFC®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR