Thallium standard solution, 1000 mg/L Tl in 0,5 M nitric acid (from TlNO₃) Certipur® standard for AAS, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.19801.0100 1.19801.0500
1.19801.0100EA 45.2 GBP
1.19801.0100 1.19801.0500
Thallium standard solution, 1000 mg/L Tl in 0,5 M nitric acid (from TlNO₃) Certipur® standard for AAS, Supelco®
Standards Cationic Standards Thallium Standards
Certifications: Traceable to SRM from NIST.