Test strips, ammonium, Reflectoquant®, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.16892.0001 1.16899.0001 1.16977.0001
1.16892.0001EA 129 GBP
1.16892.0001 1.16899.0001 1.16977.0001
Test strips, ammonium, Reflectoquant®, Supelco®
Environmental Test Kits
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly