(±)-1-Methoxy-2-propanol, EMPLURA®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

1.16738.6025 1.16738.9190 1.16738.1000
1.16738.6025EA 908 GBP
1.16738.6025 1.16738.9190 1.16738.1000
(±)-1-Methoxy-2-propanol, EMPLURA®, Supelco®
(±)-1-Methoxy-2-propanol
Formula: CH₃CH(OH)CH₂OCH₃
Boiling Pt: 119.6 °C (1013 hPa)
Melting Pt: –97 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR