TLC plates, silica gel 60, modified layers, glass backed
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.05746.0001 1.05747.0001 1.15388.0001 1.15389.0001 1.15423.0001 1.15424.0001 1.15683.0001 1.15684.0001 1.15685.0001
1.05746.0001EA 438 GBP
1.05746.0001 1.05747.0001 1.15388.0001 1.15389.0001 1.15423.0001 1.15424.0001 1.15683.0001 1.15684.0001 1.15685.0001
TLC plates, silica gel 60, modified layers, glass backed
TLC Plates
RP-2, RP-8 and RP-18 are based on silica gel 60 modified with aliphatic hydrocarbons. The chain length, in combination with the degree of modification, defines the ability to tolerate water in the solvent system and strongly affects retention. Migration time increases in the order RP-2, RP-8 and RP-18 using the same solvent composition. RP-8 and RP-18 can be run with up to 60% water in the solvent system.
  • Results less dependent on atmospheric humidity
  • Allows the use of aqueous solvent systems
  • RP-modified silica provides a ready correlation with HPLC