Kjeldahl tablets

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.10958.0250 1.10958.1000 1.15348.0250 1.15348.1000 1.17958.0250 1.16469.0250 1.16469.1000 1.18348.0250 1.18469.0250
1.10958.0250EA 203 GBP
1.10958.0250 1.10958.1000 1.15348.0250 1.15348.1000 1.17958.0250 1.16469.0250 1.16469.1000 1.18348.0250 1.18469.0250
Kjeldahl tablets
Digestion Systems Kjeldahl Tablets and Reagents

In addition to the traditional 5 g Kjeldahl tablets, which are added as catalysts to the digestion mixture, Merck Millipore can offer the 3,5 g tablet. This gives the flexibility needed to find the optimal setting for the catalyst/sulphuric acid ratio.


  • The 3,5 g tablets are suitable for microdigestion
  • Easy to use PE packaging concept

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR