Methanol, Prepsolv® for preparative chromatography, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

M-Clarity™ quality level = MQ200

1.13351.2500 1.13351.4000 1.13351.9030
1.13351.2500EA 84.3 GBP
1.13351.2500 1.13351.4000 1.13351.9030
Methanol, Prepsolv® for preparative chromatography, Supelco®
Methanol
Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR