Tungsten, powder, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

1.12406.0100
1.12406.0100EA 163 GBP
1.12406.0100
Tungsten, powder, Sigma-Aldrich®
Tungsten
Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 3370 °C
Molar Mass 183.85 g/mol
Bulk Density 4000 kg/m3
Boiling Point 5930 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR