Vanadium(III) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

1.12393.0025 1.12393.1000
1.12393.0025EA 118 GBP
1.12393.0025 1.12393.1000
Vanadium(III) chloride, Sigma-Aldrich®
Vanadium (III) chloride
Formula: Cl₃V
Density: 3 g/cm³ (20 °C)
MDL Number: MFCD00011454
CAS Number: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 157.3 g/mol
Bulk Density 1000 kg/m3
Density 3.0 g/cm3 (20 °C)
Synonyms Vanadium trichloride
Hill Formula Cl3V
Chemical Formula VCl3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR