Bioindicator, Sterikon® plus
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.10274.0001 1.10274.0002
1.10274.0001EA 94 GBP
1.10274.0001 1.10274.0002
Bioindicator, Sterikon® plus
Sterilisation Indicators
Sterikon® plus bioindicators can be used to test the functionality of autoclaves (15 minutes at 121 °C). They can also be used to check any sort of autoclave load for the success of sterilisation. For example, pharmaceuticals, particularly products in ampoules, canned food and more.
  • Easy interpretation due to colour change
  • Safe, because spores of non-pathogenic organisms (Geobacillus stearothermophilus) are used
Acc. to the recommendation of USP and EP for the use of bioindicators for checking the autoclaving of pharmaceutical products.