Ethylene glycol ≥99.5% (by GC), EMSURE® Reag. Ph. Eur., Reag. USP for analysis

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ300

1.09621.1000 1.09621.2500 1.09621.4000 1.09621.9028
1.09621.1000EA 63.9 GBP
1.09621.1000 1.09621.2500 1.09621.4000 1.09621.9028
Ethylene glycol ≥99.5% (by GC), EMSURE® Reag. Ph. Eur., Reag. USP for analysis
Ethylene glycol

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formula: HOCH₂CH₂OH
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Colour ≤ 10 Hazen
Acidity (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/20 °C) 1.113 - 1.115
Density (d 25 °C/25 °C) 1.10 - 1.12
Refractive index (n 20/D) 1.431 - 1.433
Melting point -14 - -10 °C
Boiling point 196.5 - 198.5 °C
Chloride (Cl) ≤ 0.00002 %
Fe (Iron) ≤ 0.00005 %
Formaldehyde ≤ 0.005 %
Substances reducing potassium permanganate (as O) ≤ 0.0003 %
Readily carbonizable substances conforms
Sulfated ash ≤ 0.005 %
Water ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR