pH indicator papers, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.09560.0003 1.09562.0003 1.09564.0003
1.09560.0003EA 59.2 GBP
1.09560.0003 1.09562.0003 1.09564.0003
pH indicator papers, Supelco®
Test Papers pH Indicator Papers and Test Strips
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available