Buffer solutions, Certipur®, colour coded, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
1.09475.0500EA 29.59 GBP
1.09475.0500 1.09475.9010 1.09476.0500 1.09476.9010 1.09477.0500 1.09477.9010 1.09400.0500 1.09400.9010 1.09400.4000 1.09475.4000 1.09476.4000 1.09477.4000
Buffer solutions, Certipur®, colour coded, Supelco®
Buffers pH Calibration Buffers

Materials for the precise calibration and monitoring of pH meters.


  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C


Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR