Iodine monochloride solution 0.1 mol/l acc. Wijs, Titripur® for determination of iodine value, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

1.09163.1000 1.09163.2500
1.09163.1000EA 89 GBP
1.09163.1000 1.09163.2500
Iodine monochloride solution 0.1 mol/l acc. Wijs, Titripur® for determination of iodine value, Supelco®
Iodine monochloride
Formula: ClI
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00011354
CAS Number: 7790-99-0
UN: 2789
ADR: 8 (3),II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR