Potassium permanganate 0.02 mol/l (0.1 N) aqueous solution, Titripur® Volumetric, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Permanganic acid potassium salt

MQ100

1.09121.1000 1.09121.1000_N
1.09121.1000EA 30.81 GBP
1.09121.1000 MERC1.09121.1000_N
Potassium permanganate 0.02 mol/l (0.1 N) aqueous solution, Titripur® Volumetric, Supelco®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR