Hydrogen peroxide 30% stabilised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

1.08597.1000 1.08597.2500 1.08597.9025 1.08597.9025_B 1.08597.9950
1.08597.1000EA 60.6 GBP
1.08597.1000 1.08597.2500 1.08597.9025 MERP1.08597.9025_B 1.08597.9950
Hydrogen peroxide 30% stabilised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Hydrogen peroxide

Contains 0,015 % Na₂H₂P₂O₇, 0,01 % H₃PO₄, 0,006 % NH₄NO₃ as preservatives.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Corresponds to Ph. Eur., BP, USP.

Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

Assay (manganometric) 29.0 - 31.0 %
Identity passes test
Acidity (according to Ph Eur) passes test
Acidity (according to USP) passes test
Chloride (Cl) ≤ 0.005 %
Phosphate (PO₄) ≤ 0.05 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.0003 %
Cu (Copper) ≤ 0.002 %
Pb (Lead) ≤ 0.001 %
Pd (Palladium)* ≤ 0.0001 %
Pt (Platinum)* ≤ 0.0001 %
Zn (Zinc) ≤ 0.002 %
Residual solvents (ICH Q3C) ...excluded by production process
Organic stabilizers ≤ 0.045 %
Non volatile matter (105 °C) ≤ 0.018 %
Preservatives 0.015% Na₂H₂P₂O₇; 0.01% H₃PO₄; 0.006% NH₄NO₃


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR