Titriplex® III ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

M-Clarity™ quality level = MQ100

1.08418.0100 1.08418.0250 1.08418.1000 1.08418.5000 1.08418.9025
1.08418.0100EA 54.9 GBP
1.08418.0100 1.08418.0250 1.08418.1000 1.08418.5000 1.08418.9025
Titriplex® III ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 100 g/l (20°C)
Melting Point 110 °C Elimination of water of crystallisation
Molar Mass 700 kg/m3
Bulk Density 700 kg/m3
pH 4 - 5 (50 g/l, H2O, 20 °C)
Grade ACS,ISO,Reag. Ph Eur
Synonyms Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt
Hill Formula C10H14N2Na2O8* 2 H2O
Chemical Formula C10H14N2Na2O8* 2 H2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR