Cyclopentyl methyl ether (CPME), EMPLURA®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

1.08293.1000 1.08293.2500 1.08293.4000
1.08293.1000EA 79.8 GBP
1.08293.1000 1.08293.2500 1.08293.4000
Cyclopentyl methyl ether (CPME), EMPLURA®
Cyclopentyl methyl ether (CPME)

Cyclopentyl methyl ether is a safer substitute for tetrahydrofuran, tert-butyl methyl ether, 1,4 dioxane and other ether solvents. It is produced by a 100% atomic catalytic reaction without any formation of by-products.


  • Resistance to peroxide formation improves laboratory safety
  • One-step reaction saves energy and reduces wastewater
  • More stable than tetrahydrofuran
  • Higher hydrophobicity increases yields and selectivity
  • Limited miscibility in water reduces waste stream

Formula: C₆H₁₂O
Melting Pt: –140 °C
Density: 0.86 g/cm3 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR