Silica gels for TLC and PLC plates, standardised
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.07730.1000 1.07730.5000 1.07730.9025 1.07731.1000 1.07731.5000 1.07731.9025 1.07736.1000 1.07736.2500 1.07736.9025 1.07739.1000 1.07747.1000 1.07747.2500 1.07748.1000 1.07749.1000 1.07749.2500 1.07749.9025 1.07748.2500 1.07739.9025
1.07730.1000EA 140 GBP
1.07730.1000 1.07730.5000 1.07730.9025 1.07731.1000 1.07731.5000 1.07731.9025 1.07736.1000 1.07736.2500 1.07736.9025 1.07739.1000 1.07747.1000 1.07747.2500 1.07748.1000 1.07749.1000 1.07749.2500 1.07749.9025 1.07748.2500 1.07739.9025
Silica gels for TLC and PLC plates, standardised
Chromatography Sorbents TLC Sorbents
Selection of standardised and modified silica gels that are available in many granulations and supplied from one single source, dedicated to chromatography. Standardised silica gels are widely used in separation processes to purify valuable compounds in ton quantities.
  • Reliable results and easy sample application
  • Variety of packing sizes from 1 to 25 kg
  • Economic alternatives to conventional types
* F254: Green fluorescent indicator, H: Without foreign binder, G: With gypsum, P: For preparative work.
Packaging: Supplied in plastic or tin packaging.