L-Phenylalanine, Millipore®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

M-Clarity™ quality level = MQ100

1.07256.0100 1.07256.1000 1.07256.9010
1.07256.0100EA 107 GBP
1.07256.0100 1.07256.1000 1.07256.9010
L-Phenylalanine, Millipore®
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR