Hydrogen peroxide 30%, (Perhydrol®) EMSURE® ISO for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ300

1.07209.0250 1.07209.1000 1.07209.2500 1.07209.0500
1.07209.0250EA 31.8 GBP
1.07209.0250 1.07209.1000 1.07209.2500 1.07209.0500
Hydrogen peroxide 30%, (Perhydrol®) EMSURE® ISO for analysis, Supelco®
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: –25.7 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

Assay (Manganometric) 30% min.
Color 10 Hazen max.
Other specifications on request 107 °C (1013 hPa)
Vapor Pressure 18 hPa (20 °C)
Density 1.11 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)
Grade ACS,ISO
Synonyms Perhydrol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR