n-Pentane ∼ 95%, EMPLURA®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Pentane

M-Clarity™ quality level = MQ200

1.07176.1000 1.07176.9190 1.07176.2500
1.07176.1000EA 53.5 GBP
1.07176.1000 1.07176.9190 1.07176.2500
n-Pentane ∼ 95%, EMPLURA®, Supelco®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 95.0 %
Identity (IR) conforms
Free acid (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/ 4 °C) 0.626 - 0.628
Readily carbonizable substances conforms
Evaporation residue ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR