Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® ESSENTIAL USP, Ph. Eur., BP suitable for use as excipient, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Saline, NaCl, Sea salt, Halite

1.06400.5000 1.06400.1000 1.06400.9029 1.06400.9500 1.06400.9024 1.06400.9051
1.06400.5000EA 89.2 GBP
1.06400.5000 1.06400.1000 1.06400.9029 1.06400.9500 1.06400.9024 1.06400.9051
Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® ESSENTIAL USP, Ph. Eur., BP suitable for use as excipient, SAFC®
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay (argentometric; calculated on dried substance) 99.5 - 100.5 %
Identity passes test
Appearance of solution passes test
Acidity or alkalinity passes test
Bromide (Br) ≤ 0.005 %
Ferrocyanides passes test
Iodide (I) passes test
Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water) ≤ 0.01
Phosphate (PO₄) ≤ 0.0025 %
Sulfate (SO₄) ≤ 0.0200 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Ba (Barium) passes test
Ca (Calcium) ≤ 0.002 %
K (Potassium) ≤ 0.01 %
Fe (Iron) ≤ 0.0002 %
Magnesium and alkaline-earth metals (as Ca) ≤ 0.0100 %
Residual solvents (ICH Q3C) excluded by the manufacturing process
Loss on drying (105 °C, 2 h) ≤ 0.5 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, BP, USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR