Sodium dihydrogen phosphate dihydrate, EMPROVE® ESSENTIAL JPE, BP, E339, Ph. Eur., USP suitable for use as excipient, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

1.06345.1000 1.06345.5000 1.06345.9026 1.06345.9011
1.06345.1000EA 60.6 GBP
1.06345.1000 1.06345.5000 1.06345.9026 1.06345.9011
Sodium dihydrogen phosphate dihydrate, EMPROVE® ESSENTIAL JPE, BP, E339, Ph. Eur., USP suitable for use as excipient, SAFC®
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR