Mercury(II) iodide, red Ph. Franç., extra pure, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Mercury diiodide, Mercuric iodide

M-Clarity™ quality level = MQ200

1.04420.0100 1.04420.1000
1.04420.0100EA 171 GBP
1.04420.0100 1.04420.1000
Mercury(II) iodide, red Ph. Franç., extra pure, Supelco®
Mercury (II) iodide
Formula: HgI₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 259 °C
Density: 6.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1,II
Merck Index: 13,05905

Order Now

Specification Test Results

Solubility 0.06 g/l (25 °C)
Melting Point 259 °C
Molar Mass 454.39 g/mol
Bulk Density 1350 kg/m3
Boiling Point 354 °C (1013 hPa) (sublimed)
Vapor Pressure 0.006 hPa (80 °C)
Density 6.36 g/cm3 (25 °C)
pH 6 - 7 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR