n-Hexane, EMSURE® ACS for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

M-Clarity™ quality level = MQ300

1.04367.1000 1.04367.2500 1.04367.6025 1.04367.9190 1.04367.2511
1.04367.1000EA 48.6 GBP
1.04367.1000 1.04367.2500 1.04367.6025 1.04367.9190 1.04367.2511
n-Hexane, EMSURE® ACS for analysis, Supelco®
n-Hexane

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.0 %
Identity (IR) conforms
Color ≤ 10 Hazen
Alkalinity ≤ 0.0002 meq/g
Acidity ≤ 0.0002 meq/g
Density (d 20 °C/ 4 °C) 0.659 - 0.662
Thiophene conforms
Aromatics (as benzene) ≤ 0.01 %
Sulfur compounds (as S) ≤ 0.005 %
Matter discolored by H₂SO₄ ≤ 10 Hazen
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.005 %
ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR