Guanidinium hydrochloride, Millipore®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

M-Clarity™ quality level = MQ100

1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
1.04220.0100EA 66 GBP
1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
Guanidinium hydrochloride, Millipore®
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR