Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

M-Clarity™ quality level = MQ300

1.03943.0250 1.03943.1000 1.03943.9025
1.03943.0250EA 59.75 GBP
1.03943.0250 1.03943.1000 1.03943.9025
Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
Boiling Pt: 280 - 285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1,82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (iodometric, FeCl₃·6H₂O) 99.0 - 102.0 %
Identity passes test
Insoluble matter ≤ 0.01 %
Free chlorine passes test
Acidic substances passes test
Nitrate (NO₃) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.01 %
Total nitrogen (N) ≤ 0.001 %
Total phosphorus (as PO₄) ≤ 0.01 %
Heavy metals (as Pb) ≤ 0.005 %
Ca (Calcium) ≤ 0.01 %
Cu (Copper) ≤ 0.003 %
Fe II (Iron II) ≤ 0.002 % (The Fe(II)-content increases if the recommended storage conditions (+15 to +25 °C) are not observed)
K (Potassium) ≤ 0.005 %
Mg (Magnesium) ≤ 0.005 %
Na (Sodium) ≤ 0.05 %
Zn (Zinc) ≤ 0.003 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR