Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

1.03771.2500 1.03771.9180
1.03771.2500EA 121 GBP
1.03771.2500 1.03771.9180
Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK, 1% IPA and 10 ppm Bitrex®)
Ethanol denatured

Bitrex = Denatonium benzoate

Formula: H₃CCH₂OH
Density: 0.79 g/cm³(20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR