1,2-Dichlorobenzene, EMSURE® analysis_for, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: o-Dichlorobenzene

MQ300

1.02930.1000 1.02930.2500
1.02930.1000EA 84 GBP
1.02930.1000 1.02930.2500
1,2-Dichlorobenzene, EMSURE® analysis_for, Supelco®
1,2-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 180.5
Melting Pt: –17 °C
Density: 1.309 g/cm³ (20 °C)
Flash Pt: 66
Storage Temperature: Ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1 III
Merck Index: 13,03081

Order Now

Specification Test Results

Solubility 0.13 g/l (20 °C)
Melting Point -17 °C
Molar Mass 147 g/mol
Boiling Point 180 °C (1013 hPa)
Vapor Pressure 1.33 hPa (20 °C)
Flash Point 66 °C
Explosion Limit 2.2 - 12 %(V)
Density 1.31 g/cm3 (20 °C)
Ignition Point 640 °C
Evaporation number 57


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR