Cesium nitrate ≥99%, EMSURE® for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ300

1.02856.0025 1.02856.1000
1.02856.0025EA 159 GBP
1.02856.0025 1.02856.1000
Cesium nitrate ≥99%, EMSURE® for analysis, Supelco®
Cesium nitrate
Formula: CsNO₃
Boiling Pt: 737 °C (1013 hPa)
Melting Pt: 414 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1,III
Merck Index: 12,02057

Order Now

Specification Test Results

Assay (acidimetric after ion exchange) ≥ 99 %
Chloride (Cl) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR