Calcium chloride, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

M-Clarity™ quality level = MQ100

1.02083.0250 1.02083.1000
1.02083.0250EA 424 GBP
1.02083.0250 1.02083.1000
Calcium chloride, Supelco®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Solubility 740 g/l (20 °C)
Melting Point 772 °C
Molar Mass 110.98 g/mol
Boiling Point 1600 °C
Density 2.15 g/cm3 (20 °C)
pH 8 - 10 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR