Magnesium chloride hexahydrate, crystals Ph. Eur., JPC, USP, FCC, E511, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.01872.9025
1.01872.9025EA 0 GBP
1.01872.9025
Magnesium chloride hexahydrate, crystals Ph. Eur., JPC, USP, FCC, E511, SAFC®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR