Petroleum spirit, 100…140 °C, EMPLURA® (naphtha benzine), Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

1.01770.1000 1.01770.6025 1.01770.2500
1.01770.1000EA 68.1 GBP
1.01770.1000 1.01770.6025 1.01770.2500
Petroleum spirit, 100…140 °C, EMPLURA® (naphtha benzine), Supelco®
Petroleum spirit
Boiling Pt: 100…140 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.728 g/cm³ (15 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Free acid (as CH₃COOH) ≤ 0.0005 %
Boiling range (lower value) ≥ 100.0 °C
Boiling range (upper value) ≤ 140.0 °C
Density (d 20 °C/ 4 °C) 0.720 - 0.733
Aromatics (as toluol) ≤ 0.05 %
Readily carbonizable substances conforms
Evaporation residue ≤ 0.005 %
Water ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR