Canada balsam for microscopy, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ400

1.01691.0025 1.01691.0100
1.01691.0025EA 71 GBP
1.01691.0025 1.01691.0100
Canada balsam for microscopy, Sigma-Aldrich®
Canada balsam

Canada balsam is a commonly used mounting medium to prepare permanent slides for microscopy. It is produced from the resin of the balsam fir tree and its use can be combined with xylene-containing specimens. (Refractive index (20°C) 1.515 - 1.530) Canada balsam is an IVD product and CE registered.


Analysis Note
Appearance: nearly clear, yellow resinous
Refractive index (n 20/D): 1.515 - 1.530
Turbidity: ≤ 2 NTU
Acid value: 75 - 90
Loss on drying (120 °C): ≤ 35 %
Suitability for microscopy: passes test

Density: 0.915 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00132800
CAS Number: 8007-47-4
EINECS: 232-362-2
UN: 1993
ADR: 3,III
Merck Index: 13,00948
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR