Silver sulphate ACS analytical reagent, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Disilver sulphate

M-Clarity™ quality level = MQ100

1.01509.0025 1.01509.0100 1.01509.1000
1.01509.0025EA 189 GBP
1.01509.0025 1.01509.0100 1.01509.1000
Silver sulphate ACS analytical reagent, Supelco®
Silver sulphate
Formula: Ag₂SO₄
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9,III
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 8 g/l (25 °C)
Melting Point 655 °C
Molar Mass 311.8 g/mol
Bulk Density 1200 kg/m3
Density 5.45 g/cm3 (20 °C)
pH 5 - 6 (5 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR