Ammonium nitrate, EMSURE® ACS analysis_for, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


MQ300

1.01188.0500 1.01188.1000 1.01188.5000
1.01188.0500EA 127 GBP
1.01188.0500 1.01188.1000 1.01188.5000
Ammonium nitrate, EMSURE® ACS analysis_for, Supelco®
Ammonium nitrate
Formula: NH₄NO₃
MW: 80.04 g/mol
Boiling Pt: 230
Melting Pt: 169
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1 III
Merck Index: 13,00538

Order Now

Specification Test Results

Solubility 1920 g/l (20 °C)
Melting Point 169 °C
Molar Mass 80.04 g/mol
Bulk Density 600 - 700 kg/m3
Boiling Point 302 °C (1013 hPa)
Density 1.72 g/cm3 (20 °C)
pH 5.0 - 5.5 (1920 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR