L-Alanine, Millipore®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

1.01007.0025 1.01007.0100 1.01007.9010 1.01007.1000
1.01007.0025EA 52.5 GBP
1.01007.0025 1.01007.0100 1.01007.9010 1.01007.1000
L-Alanine, Millipore®
L-Alanine

Soluble in water and mineral acids; insoluble in organic solvents.

Formula: C₃H₇NO₂
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR