1-Propanol, EMPLURA®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

MQ200

1.00996.1000 1.00996.2500 1.00996.6025
1.00996.1000EA 57 GBP
1.00996.1000 1.00996.2500 1.00996.6025
1-Propanol, EMPLURA®, Supelco®
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR